Order Entry
Denmark
ContactUsLinkComponent
 
Anti-CRMP1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
  USBI034238-PE
undefined
Anti-CRMP1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
  USBI034238-PE

 

  • Antistof type:
    Primary
  • Antigen name:
    collapsin response mediator Protein 1
  • Antigen symbol:
    CRMP1
  • Clonality:
    Polyclonal
  • Konjugation:
    PE (Phycoerythrin)
  • ELISA:
    Yes
  • Vært:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitop:
    N-terminal
  • Form:
    liquid
  • Aminosyretal:
    81 to 110
  • Storage buffer:
    PBS, pH 7,2
  • Opbevaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Størrelse:
    200 µl
  • Pakn:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: CRMP1 (collapsin response mediator protein 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human