Anti-C4BPA Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Katalog # USBI032975-FITC
Leverandør: US Biological
Specifikationer
- Antistof type:Primary
- Antigen name:complement component 4 binding Protein, alpha
- Antigen symbol:C4BPA
- Clonality:Polyclonal
- Konjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flowcytometri:Yes
- Vært:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitop:C-terminal
- Form:liquid
- Aminosyretal:477 to 507
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Størrelse:200 µl
- Pakn:200 µl
Specifikationer
Om denne vare
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: C4BPA (complement component 4 binding protein, alpha)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human