Anti-PRKDC Mouse Monoclonal Antibody [clone: 13B645]
Catalog # USBI030567
Supplier: US Biological
Specifications
- Antistof type:Primary
- Antigen name:Protein Kinase, DNA-Activated, Catalytic Polypeptide
- Antigen symbol:PRKDC
- Clonality:Monoclonal
- Clone:13B645
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Udformning:ascites
- Storage buffer:0,03% sodium azide.
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Størrelse:100 µl
- Pakn:100 µl
Specifications
About this item
Type: Primary
Antigen: PRKDC (protein kinase, DNA-activated, catalytic polypeptide)
Clonality: Monoclonal
Clone: 13B645
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human