Anti-PIK3R1 Mouse Monoclonal Antibody [clone: 13B624]
USBI030546
- Antistof type:Primary
- Antigen name:Phosphoinositide-3-kinase regulatory subunit 1
- Antigen symbol:PIK3R1
- Clonality:Monoclonal
- Clone:13B624
- Konjugation:Unconjugated
- ELISA:Yes
- Flowcytometri:Yes
- Vært:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Storage buffer:PBSwith 0,05% sodium azide and 0,5% prote
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Purification:purified
- Størrelse:100 µl
- Pakn:100 µl
Type: Primary
Antigen: PIK3R1 (phosphoinositide-3-kinase, regulatory subunit 1 (alpha))
Clonality: Monoclonal
Clone: 13B624
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse