Anti-PDK1 Mouse Monoclonal Antibody [clone: 13B613]
Catalog # USBI030535
Supplier: US Biological
Specifications
- Antistof type:Primary
- Antigen name:pyruvate dehydrogenase kinase, isozyme 1
- Antigen symbol:PDK1
- Clonality:Monoclonal
- Clone:13B613
- Konjugation:Unconjugated
- ELISA:Yes
- Flowcytometri:Yes
- Vært:Mouse
- Isotype:IgG1
- Reactivity:Monkey,Human,Rat
- Western blot:Yes
- Udformning:ascites
- Storage buffer:0,03% sodium azide.
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Størrelse:100 µl
- Pakn:100 µl
Specifications
About this item
Type: Primary
Antigen: PDK1 (pyruvate dehydrogenase kinase, isozyme 1)
Clonality: Monoclonal
Clone: 13B613
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Rat