Anti-SLC25A3/PHC Mouse Polyclonal Antibody
Product Details & Documents
Mouse Polyclonal SLC25A3/PHC antibody. Carrier free. Suitable for WB and reacts with Recombinant full length protein - Human samples. Cited in 1 publication. Immunogen corresponding to Recombinant Full Length Protein corresponding to Human SLC25A3.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: SLC25A3/PHC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Specifikationer
- Seq:1202
- Størrelse:50 µg
- Pakning:Vial
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Protein G Affinity purification
- Konjugation:Unconjugated
- Form:Liquid
- Isotype:IgG
- Antigen synonymer:OK/SW-cl.48|SLC25A3|Solute carrier family 25 member 3|Phosphate transport protein|PTP
- Koncentration:0,5 mg/ml
- Shipping temperature:4 °C
- Antigen name:PHC
- Cat. No.:ABCAAB67121-50
- Reactivity:Human
- Molekylvægt:40095 Da
- Immunogen:Recombinant Full Length Protein corresponding to Human SLC25A3
- Vært:Mouse
- Antistof type:Primary
- Opbevaringstemperatur:Upon delivery aliquot and store at −20 or −80 °C. Avoid repeated freeze/thaw cycles.
- Antigen symbol:SLC25A3/PHC
- Storage buffer:PBS



