Transporting cart
Laboratory cart, stainless steel, 886×483×924 mm, 25,4 kg
Catalog # LBCP8019000
Supplier: LABCONCO
Gå til:
Product Details & Documents
Tranporting cart made of stainless steel with two removable polished type 304 stainless steel pans that are 51 mm deep on three sides with one open side for easy equipment transfer from countertop to cart holds a variety of laboratory supplies, equipment and miscellaneous containers.
Specifikationer
- Beskrivelse:Laboratory cart, stainless steel
- B×D×H:886×483×924 mm
- Vægt:25,4 kg