Equipment table
Mobile equipment table with polyurethane castors, 1219×762×730 mm, 113,4 kg
Katalog # LBCP3819100
Leverandør: LABCONCO
Gå til:
Product Details & Documents
This mobile equipment table may be used to transport heavy equipment and to provide auxiliary bench space. With height adjustments from 28,75 to 35,75", it may be raised to the same height as most laboratory benchtops. Solid epoxy work surface, powder-coated square tubular steel frame.
- Non-marking polyurethane castors
- With toe locks
Specifikationer
- Beskrivelse:Mobile equipment table with polyurethane castors
- B×D×H:1219×762×730 mm
- Vægt:113,4 kg