Earplug dispenser, uvex one2click
Vægmonterede ørepropper dispenser 'one2click', til 600 par engangs uvex ørepropper, grå/blå
Katalog # 114-7332
Leverandør: UVEX
Gå til:
Product Details & Documents
Robust wall-mounted dispenser for 600 pairs of disposable earplugs. Installed at a convenient location, the dispenser has sufficient capacity to supply large production areas with hearing protection.
- Easy installation and a controlled release mechanism which ensures the correct number of plugs are dispensed
- Audible 'one2click' system prevents over supply thus reducing wastage costs
- Transparent design to monitor the number of earplugs remaining
- Collecting tray for catching the earplugs
Bestillingsoplysninger: Supplied without earplugs which need to be ordered separately.
Specifikationer
- Farve:grå/blå
- Beskrivelse:Vægmonterede ørepropper dispenser 'one2click', til 600 par engangs uvex ørepropper
- Cat. No.:114-7332
- Dispenser type:Kan monteres på væggen
- PPE type:Ørepropper
Accessories
Produktinformation
- Katalog #FarveTilgængelighedPrice
- Earplugs Com4-Fit, SNR 33 dB, refill box for dispenser "one 2 click" (300 pairs per refill box)Earplugs Com4-Fit, SNR 33 dB, refill box for dispenser "one 2 click" (300 pairs per refill box)UVEX2112.023orangeLog ind for tilgængelighed1 * 300 Par - 485,00 kr
Produktinformation
- Katalog #FarveTilgængelighedPrice
- X-Fit ørepropper, refill-pose til dispenser "one 2 click"X-Fit ørepropper, refill-pose til dispenser "one 2 click"UVEX2112.003intens grønLog ind for tilgængelighed1 * 200 Par - 324,00 kr
Produktinformation
- Katalog #FarveStørrelseTilgængelighedPrice
- Replacement earplugs, refill box for dispenser 'one2click' (400 pairs loose in box)Replacement earplugs, refill box for dispenser 'one2click' (400 pairs loose in box)111-1691lysegrønU*Log ind for tilgængelighed1 * 400 Par - 647,00 kr