Closed cap, Dino-Lite
Catalog # IDCPZT-Z-CC1
Supplier: Dino-Lite / IDCP B.V.
Specifications
- Beskrivelse:Closed cap to protect the lens from dust, for polarising AM series (ZT models)
- Seq:03
- Environmentally Preferable:
- Pakn:1
Specifications
About this item
Closed cap addition for the polarising AM series (ZT models). Designed to help protect the lens from dust and more.
This snap-on cover fits the AM413ZT, AM4013MZT and AM7013MZT model. Not designed for use with the AD series or AM models without polarisation.