Loading rack, RotoCycler™ 72 (consumables for Qiagen Rotor-Gene™ Q)
Catalog # SIMPT319-4WS1
Supplier: Simport Scientific
Specifications
- Beskrivelse:Workstation, aluminum, for 72 tubes
- Pakn:1
Specifications
About this item
Solid aluminium loading rack for handling and insertion of caps on tubes.
Holds up to 18×0,1 ml tube strips (total of 72 tubes) and other cavities can hold larger reaction tubes. It should be simply place in the refrigerated area to keep reactions cool during setup.
All wells are numbered and colour coding is made possible by inserting a Capinsert™ in up to two locations.
Five hundred colour coding inserts of assorted colours are enclosed.