Levelling table, SP Bel-Art
Levelling table, 203×203×9,5 mm
Katalog # BELAH183100000
Leverandør: Bel-Art Products, a Part of SP
Gå til:
Product Details & Documents
This clear acrylic level surface helps when pouring immunological plates, horizontal agarose gels and acrylamide sequencing gels.
- Each leg is adjustable
- Inset levelling bulb below the working surface assists in accurate adjustment
Specifikationer
- Beskrivelse:Levelling table, 203×203×9,5 mm