Orders LinkContactUsLinkComponent
Cold traps
Cold traps
Cold trap, S 29-NW-16
Katalog # KGWA17380-NW16
Leverandør:   KGW
Cold traps
Katalog # KGWA17380-NW16
Leverandør:  KGW
Cold traps
Cold traps
Cold trap, S 29-NW-16
Price
9.270,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Simple cold traps, which are also known as cold finger condensers, are used in conjunction with vacuum pumps and the condensation Produced from humidity or solvents these cold traps can be used for many different tasks. With glass vacuum flanges you are able to connect this cold trap directly to the metal flanges of a high vacuum pump.
Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
  • Reliable, perfect version, easy to clean and handle
  • No stand material for holding the cold trap
  • Protective casing of Dewar flask made of blue coated metal or aluminum
  • Pressure-free cooling sphere inside the Dewar flask
These simple cold finger condensers are used in standard applications in conjunction with vacuum pumps and the condensation is forced out of the humidity or solvents used in the chemical applications. Cold traps can be changed very quickly without any problems and it can be connected directly to the metal flanges of a high vacuum pump with its glass vacuum flanges.
Specifikationer
  • Beskrivelse:
    Cold trap, S 29-NW-16
  • Seq:
    01
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Cold trap SL 29-NW16-A, spare part, with spout, vacuum flange KF NW 16, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 ml
    Cold trap SL 29-NW16-A, spare part, with spout, vacuum flange KF NW 16, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 ml
    KGWA17385-NW16
    Log ind for tilgængelighed
    1 * 1 Items - 3.780,00 kr