Zoology: prepared slides with protozoa
Katalog # LIED74700
Leverandør: LIEDER JOHANNES
Specifikationer
- Beskrivelse:Prepared slides with protozoa (10 pcs.)
- Environmentally Preferable:
- Pakn:1
Specifikationer
Om denne vare
These prepared slides with protozoa include:
- Rhizopoda (Amoeba proteus) w.m.
- Radiolaria, mixed species, fossil
- Foraminifera from Mediterranean sea, mixed species, recent
- Euglena viridis, Flagellata, w.m.
- Ceratium hirundinella, fresh-water Dinoflagellate
- Trypanosoma gambiense, blood smear, causing African sleeping sickness
- Plasmodium, causes human malaria, blood smear
- Eimeria stiedae, infected liver, different phases of developing, sec.
- Paramecium, micro- and macronuclei stained, a common ciliate
- Vorticella, a coloniate ciliate