Robotic tips for Hamamatsu 6000 and 7000 systems, Axygen®
Catalog # AXYGFDSS-1536-BK-R
Supplier: Corning
Specifications
- Pakket:384 tips/rack, 10 racks/pack, 5 packs/case
- Steril:-
- Volumen:50 µl
- Environmentally Preferable:Lav fremstillingspåvirkning
- Miljøcertificering:I-RECs, ISO 14001 - Reynosa
- Spits farge:Black
- Til brug med:Head FDSS 7000
- Filter:-
- Pakn:19.200
Specifications
About this item
These black automation tips made of PP are engineered to the exact specifications of the original equipment manufacturers’ tips, ensuring complete compatibility and a consistent fit.
- Conductive tips
- Certified DNase-/RNase-free and non pyrogenic for complete protection against contamination
- Optimised fine point for accurate dispensing