Orders LinkContactUsLinkComponent
Temperature sensors
Supplier:   WINKLER
Temperature sensors
Supplier:  WINKLER
Certificates
Temperature sensors
Temperature sensors
5 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Beskrivelse
    Område
    Ø×L
    Availability
    Price
  • Temperature probe, PT100 with two wire and cable (1,8 m)
    +250…+500 °C
    4×50 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Temperature probe, PT100 with four wire and cable (1,8 m)
    +250…+500 °C
    4×50 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Thermocouple tape, type J (Fe-CuNi), with wire end ferrules and cable (1,2 m)
    +500…+1000 °C
    16×450 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Thermocouple tape, type J (NiCr-Ni), with wire end ferrules and cable (1,2 m)
    +500…+1000 °C
    16×450 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Thermocouple rod, type J (Fe-CuNi) and cable (1,2 m)
    +500…+1000 °C
    1,6×300 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
Compensating cables
Product Information
  • Catalog #
    Længde
    Beskrivelse
    Farve
    Availability
    Price
  • 10 m
    Compensating cable, type J
    sort/hvid
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 0,00 kr
    0,00 kr
    1 * 1 Items
       0,00 kr
Product Information
  • Catalog #
    Længde
    Beskrivelse
    Farve
    Availability
    Price
  • 10 m
    Compensating cable, type K
    grøn/hvid
    Log in for availability
    Log in for availability
    1 * 10 m
    1 * 10 m - 0,00 kr
    0,00 kr
    1 * 10 m
       0,00 kr