Wall-mounted cabinets
Supplier: KOETTERMANN
Jump to:
About this item
Made of epoxy powder coated steel, with one shelf.
- Solid hinged doors or sliding glass doors
- Height adjustable shelfs
- Maximum loading capacity: 80 kg
31 Options Available Below
Product Details & Documents
Made of epoxy powder coated steel, with one shelf.
- Solid hinged doors or sliding glass doors
- Height adjustable shelfs
- Maximum loading capacity: 80 kg
Product Family Options
Product Information
- BeskrivelseDøreB×D×HAvailabilityPrice
- Wall-mounted cabinet, with installation cabinet1 door, left hinged300×366×630 mmLog in for availability1 * 1 Items2.430,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00033 - Wall-mounted cabinet, with installation cabinet1 door, right hinged300×366×630 mmLog in for availability1 * 1 Items2.410,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00034 - Wall-mounted cabinet, with installation cabinet1 door, right hinged300×366×780 mmLog in for availability1 * 1 Items2.590,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00036 - Wall-mounted cabinet, with installation cabinet1 door, left hinged300×366×780 mmLog in for availability1 * 1 Items2.590,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00035 - Wall-mounted cabinet, with one shelf1 door, left hinged450×366×480 mmLog in for availability1 * 1 Items2.470,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00037 - Wall-mounted cabinet, with one shelf, lockable1 door, left hinged450×366×480 mmLog in for availability1 * 1 Items2.660,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00038 - Wall-mounted cabinet, with one shelf1 door, right hinged450×366×480 mmLog in for availability1 * 1 Items2.470,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00039 - Wall-mounted cabinet, with one shelf, lockable1 door, right hinged450×366×480 mmLog in for availability1 * 1 Items2.650,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00040 - Wall-mounted cabinet, with one shelf1 door, left hinged450×366×630 mmLog in for availability1 * 1 Items2.710,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00041 - Wall-mounted cabinet, with one shelf, lockable1 door, left hinged450×366×630 mmLog in for availability1 * 1 Items2.910,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00042 - Wall-mounted cabinet, with one shelf1 door, right hinged450×366×630 mmLog in for availability1 * 1 Items2.780,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00043 - Wall-mounted cabinet, with one shelf, lockable1 door, right hinged450×366×630 mmLog in for availability1 * 1 Items2.890,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00044 - Wall-mounted cabinet, with one shelf1 door, left hinged450×366×780 mmLog in for availability1 * 1 Items2.900,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00045 - Wall-mounted cabinet, with one shelf, lockable1 door, left hinged450×366×780 mmLog in for availability1 * 1 Items3.080,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00046 - Wall-mounted cabinet, with one shelf1 door, right hinged450×366×780 mmLog in for availability1 * 1 Items2.890,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00047 - Wall-mounted cabinet, with one shelf, lockable1 door, right hinged450×366×780 mmLog in for availability1 * 1 Items3.100,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00048 - Wall- mounted cabinet, with one shelf, open front-600×350×480 mmLog in for availability1 * 1 Items1.840,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00049 - Wall- mounted cabinet, with one shelf, open front-600×350×630 mmLog in for availability1 * 1 Items1.950,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00054 - Wall- mounted cabinet, with one shelf, open front-600×350×780 mmLog in for availability1 * 1 Items2.110,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00059 - Wall-mounted cabinet, with one shelf1 door, right hinged600×366×480 mmLog in for availability1 * 1 Items2.740,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00052 - Wall-mounted cabinet, with one shelf, lockable1 door, right hinged600×366×480 mmLog in for availability1 * 1 Items2.970,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00053 - Wall-mounted cabinet, with one shelf1 door, left hinged600×366×480 mmLog in for availability1 * 1 Items2.760,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00050 - Wall-mounted cabinet, with one shelf, lockable1 door, left hinged600×366×480 mmLog in for availability1 * 1 Items2.960,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00051 - Wall-mounted cabinet, with one shelf1 door, left hinged600×366×630 mmLog in for availability1 * 1 Items2.960,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00055 - Wall-mounted cabinet, with one shelf, lockable1 door, left hinged600×366×630 mmLog in for availability1 * 1 Items3.170,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00056 - Wall-mounted cabinet, with one shelf1 door, right hinged600×366×630 mmLog in for availability1 * 1 Items2.960,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00057 - Wall-mounted cabinet, with one shelf, lockable1 door, right hinged600×366×630 mmLog in for availability1 * 1 Items3.170,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00058 - Wall-mounted cabinet, with one shelf1 door, left hinged600×366×780 mmLog in for availability1 * 1 Items3.230,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00060 - Wall-mounted cabinet, with one shelf, lockable1 door, left hinged600×366×780 mmLog in for availability1 * 1 Items3.460,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00061 - Wall-mounted cabinet, with one shelf, lockable1 door, right hinged600×366×780 mmLog in for availability1 * 1 Items3.470,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00063 - Wall-mounted cabinet, with one shelf1 door, right hinged600×366×780 mmLog in for availability1 * 1 Items3.230,00 krSpecifications:
More
Specifications
Cat. No.KOTT307-00062