Orders LinkContactUsLinkComponent
Digital block heater, compact
Digital block heater, compact
Certificates
Digital block heater, compact
2 Options Available Below
Product Details & Documents
These versatile digital block heaters provide good temperature uniformity and stability ensuring reproducible results. Units are compact, light weight with digital temperature controls and display, they maximises valuable bench space. The units are ideal for sample preparation, enzyme preservation, DNA amplification and enzyme–substrate reactions.
  • Built-in temperature-sensing probe assists with outstanding temperature accuracy and control
  • Designed for precise temperature control
  • Fast heating rate
  • Multiple combination of different size of blocks allows for maximum tube size versatility during experimental procedure
  • 12V low voltage power input
Delivery information: Supplied with block extraction tool and plastic lid, but without blocks, these must be ordered separately.
Caution: Temp. stability value shown above is at 37 °C.
* RT = Ambient
** S - 1 block, 2×½ blocks, 4×¼ blocks or 1×½ block plus 2×¼ blocks
*** D - 2 blocks, 4×½ blocks, 8×¼ blocks or combination
Specifications for Product Family
Specifications
  • Catalog No:
  • 460-0284
  • 460-0283
  • Temperaturinterval:
  • RT* +2...100 °C
  • RT* +2...100 °C
  • Temperaturstabilitet:
  • ≤±0,5 °C
  • ≤±0,5 °C
Specifications
Product Family Options
Product Information
  • Model
    Antal blokke
    Opvarmnings kapacitet
    Temperaturområde
    Temp. stabilitet
    B×D×H
    Vægt
    Availability
    Price
  • Compact Dry Bath S**
    1
    58 W
    RT* +2...100 °C
    ≤±0,5 °C
    185×185×25 mm
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.990,00 kr
    2.990,00 kr
    1 * 1 Items
       2.990,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    460-0284
    Temperaturinterval
    RT* +2...100 °C
    Temperaturstabilitet
    ≤±0,5 °C
  • Compact Dry Bath D***
    2
    77 W
    RT* +2...100 °C
    ≤±0,5 °C
    200×200×25 mm
    1,5 kg
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 4.030,00 kr
    4.030,00 kr
    1 * 1 Items
       4.030,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    460-0283
    Temperaturinterval
    RT* +2...100 °C
    Temperaturstabilitet
    ≤±0,5 °C
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Til
    Ant. huller
    Availability
    Price
  • 460-0318
    ½ aluminium block
    50 ml tubes
    3
    Log in for availability
    1 * 1 Items - 641,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Ant. huller
    Availability
    Price
  • 460-0315
    ¼ aluminium block
    1,5 ml tubes
    6
    Log in for availability
    1 * 1 Items - 185,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Ant. huller
    Availability
    Price
  • 460-0316
    ¼ aluminium block
    2,0 ml tubes
    6
    Log in for availability
    1 * 1 Items - 185,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Ant. huller
    Availability
    Price
  • 460-0317
    ½ aluminium block
    15 ml tubes
    6
    Log in for availability
    1 * 1 Items - 641,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Ant. huller
    Availability
    Price
  • 460-0323
    Plastic lid
    Compact D models
    -
    Log in for availability
    1 * 1 Items - 117,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Ant. huller
    Availability
    Price
  • 460-0322
    Plastic lid
    Compact S models
    -
    Log in for availability
    1 * 1 Items - 70,90 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Ant. huller
    Availability
    Price
  • 460-0321
    Heat insulator
    Compact D models
    -
    Log in for availability
    1 * 1 Items - 151,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    B×D×H
    Availability
    Price
  • 460-0319
    Full aluminium block
    96-well microplate
    108×72×18 mm
    Log in for availability
    1 * 1 Items - 806,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Ant. huller
    Availability
    Price
  • 460-0320
    Heat insulator
    Compact S models
    -
    Log in for availability
    1 * 1 Items - 92,60 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Ant. huller
    Availability
    Price
  • 460-0314
    ¼ aluminium block
    0,5 ml tubes
    12
    Log in for availability
    1 * 1 Items - 209,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Ant. huller
    Availability
    Price
  • 460-0313
    ¼ aluminium block
    0,2 ml Ø tubes
    24
    Log in for availability
    1 * 1 Items - 232,00 kr