Orders LinkContactUsLinkComponent
Media set
Media set
Total bacteria media set, Total Count, 3 Tests
Catalog # HACH2277700
Supplier:   Hach
Media set
Catalog # HACH2277700
Supplier:  Hach
CAS Number:  
Media set
Media set
Total bacteria media set, Total Count, 3 Tests
CAS Number:  
Price
553,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
Media set is a colorimetric method that is used for determination of total bacteria.
  • ToxTrak™ Toxicity test speeds sample evaluations
  • Economically alternative to bioassays
It includes dehydrated tryptic soy broth, screw cap vial and sterile transfer pippets.
Specifications
Specifications
  • Pakn:
    3 Tests
  • Beskrivelse:
    Total bacteria media set
  • Til:
    Total Count
Specifications