Orders LinkContactUsLinkComponent
ECR masking reagent solution
ECR masking reagent solution
ECR masking reagent solutionfor aluminium determination, 0,006 - 0,25 mg/l, Eriochrome cyanine R, 25 mL
Katalog # HACH2380123
Leverandør:   Hach
ECR masking reagent solution
Katalog # HACH2380123
Leverandør:  Hach
CAS nummer:  
ECR masking reagent solution
ECR masking reagent solution
ECR masking reagent solutionfor aluminium determination, 0,006 - 0,25 mg/l, Eriochrome cyanine R, 25 mL
CAS nummer:  
Price
248,00 kr
Log ind for tilgængelighed
1 * 25 mL
Product Details & Documents
This solution is used for Aluminum determination by the Eriochrome Cyanine R Method.
Documents
Specifikationer
Specifikationer
  • Kemisk metode:
    Eriochrome cyanine R
  • Pakn:
    25 mL
  • Beskrivelse:
    ECR masking reagent solutionfor aluminium determination
  • Område:
    0,006 - 0,25 mg/l
Specifikationer