Orders LinkContactUsLinkComponent
EDTA solutions
EDTA solutions
Leverandør:   Hach
EDTA solutions
Leverandør:  Hach
Certifikater
EDTA solutions
5 tilgængelige muligheder nedenfor
Product Details & Documents
Documents
Product Family Options
Produktinformation
  • Område
    Kemisk metode
    Pakning
    Pakn
    Tilgængelighed
    Price
  • HACH2241926
    EDTA solution, 1 M, 50 ml, for determination of calcium and magnesium hardness in water by the calmagite colorimetric method
    EDTA solution, 1 M, 50 ml, for determination of calcium and magnesium hardness in water by the calmagite colorimetric method
    Marked dropping bottle
    50 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 50 mL
    1 * 50 mL - 200,00 kr
    200,00 kr
    1 * 50 mL
       200,00 kr
  • HACH20553
    EDTA standard solution, 0,0200 N, 1 l, for hardness determination by buret titration
    EDTA standard solution, 0,0200 N, 1 l, for hardness determination by buret titration
    Self-contained dropping bottle
    1 L
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 L
    1 * 1 L - 314,00 kr
    314,00 kr
    1 * 1 L
       314,00 kr
  • HACH2349932
    EDTA standard solution, 0,035 N, 100 ml, for hardness determination by drop-count titration
    EDTA standard solution, 0,035 N, 100 ml, for hardness determination by drop-count titration
    100 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mL
    1 * 100 mL - 255,00 kr
    255,00 kr
    1 * 100 mL
       255,00 kr
  • HACH2268732
    Alkaline EDTA solution, 100 ml , for determination of exchangeable potassium in soils
    Alkaline EDTA solution, 100 ml , for determination of exchangeable potassium in soils
    Marked dropping bottle
    100 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mL
    1 * 100 mL - 269,00 kr
    269,00 kr
    1 * 100 mL
       269,00 kr
  • HACH2498132
    EDTA standard solution, 0,0075N, 100 ml marked dropping bottle
    EDTA standard solution, 0,0075N, 100 ml marked dropping bottle
    0 - 360 ka/ha
    Gypsum requirement drop count titration
    100 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mL
    1 * 100 mL