Orders LinkContactUsLinkComponent
Visual comparator test kits, iron
Visual comparator test kits, iron
Supplier:   Hach
Visual comparator test kits, iron
Supplier:  Hach
Certificates
Visual comparator test kits, iron
Visual comparator test kits, iron
5 Options Available Below
Product Details & Documents
Qualitative results using traditional colour comparator disc methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case for 100 tests
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Product Family Options
Product Information
  • Beskrivelse
    Område
    Kemisk metode
    Test pr. sæt
    Pakn
    Availability
    Price
  • Iron test kit, colour disc model IR-18 A
    0,02 - 1 mg/l
    1,10-Phenanthroline
    100
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.580,00 kr
    1.580,00 kr
    1 * 1 Each
       1.580,00 kr
  • Iron test kit, colour disc model IR-21
    0 - 0,1 mg/l
    0 - 1,2 mg/l
    TPTZ
    100
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 2.100,00 kr
    2.100,00 kr
    1 * 1 Each
       2.100,00 kr
  • Iron test kit, colour disc model IR-24
    0 - 1,0 mg/l
    Ferrozine
    100
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 2.130,00 kr
    2.130,00 kr
    1 * 1 Each
       2.130,00 kr
  • Iron test kit, colour disc model IR-18
    0 - 4 mg/l
    1,10-Phenanthroline
    100
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.310,00 kr
    1.310,00 kr
    1 * 1 Each
       1.310,00 kr
  • Iron test kit, colour disc model IR-18 B
    0,25 - 7 mg/l
    1,10-Phenanthroline
    100
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.310,00 kr
    1.310,00 kr
    1 * 1 Each
       1.310,00 kr