Orders LinkContactUsLinkComponent
Powder pillows, PathoScreen Medium Presence/Absence
Powder pillows, PathoScreen Medium Presence/Absence
Leverandør:   Hach
Powder pillows, PathoScreen Medium Presence/Absence
Leverandør:  Hach
Certifikater
Powder pillows, PathoScreen Medium Presence/Absence
Powder pillows, PathoScreen Medium Presence/Absence
2 tilgængelige muligheder nedenfor
Product Details & Documents
These are used to detect waterborne pathogens associated with fecal contamination of water sources.
  • Simple presence-absence method eliminates need for an incubator in tropical and semi-tropical climates
  • Complete Solutions for testing drinking water
Product Family Options
Produktinformation
  • Pakn
    Tilgængelighed
    Price
  • HACH2610696
    PathoScreen medium presence / absence powder pillows, 100 ml samples
    PathoScreen medium presence / absence powder pillows, 100 ml samples
    50
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 50 Items
    1 * 50 Items - 749,00 kr
    749,00 kr
    1 * 50 Items
       749,00 kr
  • HACH2610796
    PathoScreen medium MPN pillows, 20 ml samples
    PathoScreen medium MPN pillows, 20 ml samples
    50
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 50 Items
    1 * 50 Items - 707,00 kr
    707,00 kr
    1 * 50 Items
       707,00 kr