Orders LinkContactUsLinkComponent
Multiparameter test kits, model HA-62
Multiparameter test kits, model HA-62
Supplier:   Hach
Multiparameter test kits, model HA-62
Supplier:  Hach
Certificates
Multiparameter test kits, model HA-62
Multiparameter test kits, model HA-62
2 Options Available Below
Product Details & Documents
A multi-parameter test kit for hardness, iron and pH, which provides an easy-to-use, cost effective way to determine the quality of water.
  • Popular tests in one rugged kit
  • Uses color disc and drop count titration methods
Product Family Options
Product Information
  • Beskrivelse
    Område
    Pakn
    Availability
    Price
  • For hardness (as CaCO₃), iron and pH, HA-62
    Hardness: 17,19 - 342,36 mg/l, Iron: 0,1 - 5 mg/l, pH: 6,5 - 8,5
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 3.470,00 kr
    3.470,00 kr
    1 * 1 Each
       3.470,00 kr
  • For hardness (as CaCO₃), iron and pH, HA-62B
    Hardness: 17,19 - 342,36 mg/l, Iron: 0,2 - 10 mg/l, pH: 4 - 10
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 3.880,00 kr
    3.880,00 kr
    1 * 1 Each
       3.880,00 kr