Spill containment units
PIG® Poly IBC containment unit, 1363 litres l, 1570×1570×660 mm
Katalog # 121-0184
Leverandør: NEW PIG
Gå til:
Product Details & Documents
Spill containment units made from LDPE.
- Catch spills if containers leak, drip or crack
- Low profile for easy access to and dispensing from tanks
- Resists UV rays, rust, corrosion and most chemicals
If this product is used with the flammable liquids, please consider the regulations that apply to storage and handling of flammable liquids and the safety of this application, specifically flammable vapours, static discharge and heat sources.
Specifikationer
- Kapacitet:1363 litres l
- Beskrivelse:PIG® Poly IBC containment unit
- B×D×H:1570×1570×660 mm