Orders LinkContactUsLinkComponent
Blocking buffers
Blocking buffers
Blocking buffer for fluorescent western blotting, 500 mL
Katalog # ROCKMB-070
Leverandør:   Rockland Immunochemicals
Blocking buffers
Katalog # ROCKMB-070
CAS nummer:  
Blocking buffers
Blocking buffers
Blocking buffer for fluorescent western blotting, 500 mL
CAS nummer:  
Price
2.530,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
A blocking buffer specifically designed for western blotting using fluorochrome conjugated antibodies. Blocking buffer was prepared using ultra pure reagents dissolved in pharmaceutical grade water (WFI) and consists of a proprietary protein formulation in tris buffered saline at pH 7,6 with thimerisol added as an antimicrobial agent.
Documents
Specifikationer
Specifikationer
  • Pakn:
    500 mL
  • Beskrivelse:
    Blocking buffer for fluorescent western blotting