Orders LinkContactUsLinkComponent
Cold traps, BOLA
Cold traps, BOLA
Cold traps, PFA, collection capacity 320 ml, O.Ø×H 60×400 mm
Katalog # BOHLB317-60
Leverandør:   Bohlender
Cold traps, BOLA
Katalog # BOHLB317-60
Leverandør:  Bohlender
Cold traps, BOLA
Cold traps, BOLA
Cold traps, PFA, collection capacity 320 ml, O.Ø×H 60×400 mm
Price
14.400,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PTFE head with two threaded necks GL14 for connection of hard-walled tubing by means of separately available laboratory screw joints. Connecting nut is made of PTFE reinforced with glassfibres. GL hose connectors are recommend for soft, flexible tubing. Temperature resistant from −200 to +250 °C with suitable vaccum transparent. PFA, transparent.
Specifikationer
  • Beskrivelse:
    Cold traps, PFA, collection capacity 320 ml, O.Ø×H 60×400 mm