Classic syringes, Dosys™, WHEATON®
Katalog # WHEAW852006
Supplier: DWK Life Sciences
Specifications
- Kapacitet:0,025 - 0,3 ml
- Beskrivelse:Dosys™ 163 classic syringe
- Seq:01
- Inddeling:0,025 ml
- Pakn:1
Specifications
About this item
Fast and reliable volume setting, high chemical resistance, and excellent reproducibility make this syringe ideal for laboratories.
- Automatic refilling
- Integrated valve system
- Luer lock nozzle
- Two-ring and pistol-grip handle
Delivery: Supplied with silicone feed tube, sinker, aspiration and vent cannulas, and operating manual.