ContactUsLinkComponent
FOCUS™ protein reductant kit
FOCUS™ protein reductant kit
Katalog # 786-230
Leverandør:  G-Biosciences
CAS nummer:  
undefined
FOCUS™ protein reductant kit
Katalog # 786-230
Leverandør:  G-Biosciences
CAS nummer:  
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    FOCUS™ Protein Reductant Kit
Specifikationer
Om denne vare
FOCUS™ Protein Reductant Kit is a water soluble, odorless, non-toxic and stable TCEP [Tris (2-carboxyethyl) phosphine] reductant for protein reduction.
FOCUS™ Protein Reductant has improved stability and efficiency compared to DTT and reduces proteins over a wide range of pH, including lower acidic pH. FOCUS™ Protein Reductant completely reduces highly stable disulfide bonds in less than 5 minutes at room temperature.
FOCUS™ Protein Reductant is compatible with the alkylation of thiol groups for 2D analysis, as it does not compete with the alkylation reagent iodoacetamide, unlike DTT and other commonly used reductants.
The kit is supplied with a proprietary buffer necessary for efficient reduction of disulfide bonds, while minimising reoxidation of the competing thiol pairs in protein samples.