Orders LinkContactUsLinkComponent
Capping tool for PCR plates
Capping tool for PCR plates
Capping tool
Katalog # 731-0702
Leverandør:   Brand
Capping tool for PCR plates
Katalog # 731-0702
Leverandør:  Brand
Capping tool for PCR plates
Capping tool for PCR plates
Capping tool
Price
469,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Cap tool for attaching and detaching caps on PCR plates.
  • Ensures secure closing with little effort
  • Facilitates contamination-free removal
Specifikationer
  • Beskrivelse:
    Capping tool