Sterile anti-fog solution for autoclavable goggles, BioClean™
Catalog # 113-8145
Supplier: Nitritex
Specifications
- Beskrivelse:Sterile anti-fog solution for autoclavable goggles, BioClean, 60 ml bottle
- Størrelse:60 ml
- Pakn:10
Specifications
About this item
Sterile anti-fog spray for autoclavable goggles.
- Only two sprays to be applied on the inside of the goggles
- Excess liquid from the inside to be dried by cleanroom wipe
- To be used after each autoclave cycle
- Sterilised by gamma irradiation