Capillary tubes
Capillary Tubes, 1,50 mm, 20 µl
Katalog # SARS19.447.001
Marketsource
Leverandør: SARSTEDT
Gå til:
Product Details & Documents
Glass, transparent.
- End-to-end capillaries
- Prepared with potassium EDTA
Specifikationer
- Ø:1,50 mm
- Volumen:20 µl
- Beskrivelse:Capillary Tubes