Orders LinkContactUsLinkComponent
Inclusion bodies solubilisation buffer
Inclusion bodies solubilisation buffer
Leverandør:   G-Biosciences
Inclusion bodies solubilisation buffer
Leverandør:  G-Biosciences
Certifikater
Inclusion bodies solubilisation buffer
Inclusion bodies solubilisation buffer
2 tilgængelige muligheder nedenfor
Product Details & Documents
The IBS™ buffer is specifically developed for solubilisation of inclusion bodies.
IBS-HP™ Buffer is for the solubilisation of inclusion bodies containing highly hydrophobic proteins.
Product Family Options
Produktinformation
  • Pakn
    Tilgængelighed
    Price
  • 786-183
    IBS™ buffer
    IBS™ buffer
    100 mL
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 mL
    1 * 100 mL - 2.140,00 kr
    2.140,00 kr
    1 * 100 mL
       2.140,00 kr
  • 786-183HP
    IBS-HP™ buffer
    IBS-HP™ buffer
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 2.140,00 kr
    2.140,00 kr
    1 * 1 Each
       2.140,00 kr