Orders LinkContactUsLinkComponent
Pencil sharpener, container type
Pencil sharpener, container type
Sharpener for 8 mm pencils, 400×660 mm, 20 g, Krom/sort
Katalog # 811-2105
Leverandør:   WEDO WERNER DORSCH
Pencil sharpener, container type
Katalog # 811-2105
Leverandør:  WEDO WERNER DORSCH
Pencil sharpener, container type
Sharpener for 8 mm pencils, 400×660 mm, 20 g, Krom/sort
Price
67,80 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Plastic pencil sharpener with chrome bottom and lid.
  • For 8 mm pencils, with interchangeable replacement blade.
Specifikationer
  • Farve:
    Krom/sort
  • Beskrivelse:
    Sharpener for 8 mm pencils, 400×660 mm, 20 g