Orders LinkContactUsLinkComponent
VWR® Sample Vials
VWR® Sample Vials
VWR® Sample Vials
VWR® Sample Vials
These sample vials are made from PTFE.
  • Ideal for storing aggressive or expensive materials
  • Suitable for small scale reactions at low pressure
  • Tapered inner for easy removal of contents
  • Thick wall construction
Product Family Options
  • Kapacitet
    Diameter×H
    Kasket inkluderet
  • 25,00 ml
    33×64 mm
    Yes
    1 * 1 Items
    1 * 1 Items - 814,00 kr
    814,00 kr
    1 * 1 Items
       814,00 kr
  • 50,00 ml
    43×78 mm
    Yes
    1 * 1 Items
    1 * 1 Items - 1.020,00 kr
    1.020,00 kr
    1 * 1 Items
       1.020,00 kr