Orders LinkContactUsLinkComponent
Protease inhibitor set
Protease inhibitor set
Protease Inhibitor Set, 1 SET
Katalog # 786-207
Leverandør:   G-Biosciences
Protease inhibitor set
Katalog # 786-207
Leverandør:  G-Biosciences
CAS nummer:  
Protease inhibitor set
Protease inhibitor set
Protease Inhibitor Set, 1 SET
CAS nummer:  
Price
3.150,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Protease inhibitor set contains 12 ready to use individual protease inhibitors, each at a 100X concentration. Various concentrations and/or combinations of protease inhibitors may be used to inhibit a broad spectrum of protease activity. The Protease inhibitor set can be used to design specific protease inhibitor cocktails, supplement existing cocktails or to screen for specific protease classes.
Leveringsinformation: Set includes each of the following individual protease inhibitors: AEBSF, ALLN, Antipain dihydrochloride, Aprotinin, Bestatin, Chymostatin, E-64, EDTA-Na₂, Leupeptin, Pepstatin, Phosphoramidon, and PMSF. Individual proteases are also available as dry powders.
Documents
Specifikationer
Specifikationer
  • Pakn:
    1 SET
  • Beskrivelse:
    Protease Inhibitor Set