Orders LinkContactUsLinkComponent
Biotin kits, HOOK™
Biotin kits, HOOK™
Leverandør:   G-Biosciences
Biotin kits, HOOK™
Leverandør:  G-Biosciences
Certifikater
Biotin kits, HOOK™
18 tilgængelige muligheder nedenfor
Product Details & Documents
HOOK™ Biotin Reagent that conjugates to proteins through amines, sulfhydryls, carboxyls or carbohydrates. The amine and sulfhydryl coupling HOOK™ Biotin Reagents couple directly to the protein through their reactive groups, however the carboxyl coupling HOOK™ Biotin Reagents require a carbodiimide crosslinker and the carbohydrate coupling HOOK™ Biotin Reagents require carbohydrate oxidation before coupling.
  • Optimizer Buffer™ for improved coupling efficiency
  • SpinOUT™ gel filtration columns for rapid (<10 minutes) purification
  • Biotin assay reagents to determine level of biotin incorporation
  • Labels 1 to 10 mg protein/reaction
  • Suitable for 10 coupling reactions
The kits include EDC as the carbodiimide crosslinker in the carboxyl coupling kits and sodium meta-periodate for carbohydrate oxidation in the carbohydrate coupling kits. Kit also contains a specific Optimizer Buffer™ that provides the optimal reaction conditions for each HOOK™ Biotin Reagent.
Product Family Options
Produktinformation
  • Pakn
    Tilgængelighed
    Price
  • BS-01
    NHS-Biotin kit
    NHS-Biotin kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 2.370,00 kr
    2.370,00 kr
    1 * 1 Each
       2.370,00 kr
  • BS-02
    NHS-LC-Biotin kit
    NHS-LC-Biotin kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 3.230,00 kr
    3.230,00 kr
    1 * 1 Each
       3.230,00 kr
  • BS-03
    NHS-LC-LC-Biotin kit
    NHS-LC-LC-Biotin kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 3.760,00 kr
    3.760,00 kr
    1 * 1 Each
       3.760,00 kr
  • BS-04
    NHS-SS-Biotin kit
    NHS-SS-Biotin kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 3.930,00 kr
    3.930,00 kr
    1 * 1 Each
       3.930,00 kr
  • BS-05
    NHS-dPEG₄™-Biotin kit
    NHS-dPEG₄™-Biotin kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 3.910,00 kr
    3.910,00 kr
    1 * 1 Each
       3.910,00 kr
  • BS-06
    Sulpho-NHS-Biotin kit
    Sulpho-NHS-Biotin kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 3.790,00 kr
    3.790,00 kr
    1 * 1 Each
       3.790,00 kr
  • BS-07
    Sulpho-NHS-LC-Biotin kit
    Sulpho-NHS-LC-Biotin kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 3.710,00 kr
    3.710,00 kr
    1 * 1 Each
       3.710,00 kr
  • BS-08
    Sulpho-NHS-LC-LC-Biotin kit
    Sulpho-NHS-LC-LC-Biotin kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 3.850,00 kr
    3.850,00 kr
    1 * 1 Each
       3.850,00 kr
  • BS-09
    Sulpho-NHS-SS-Biotin kit
    Sulpho-NHS-SS-Biotin kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 3.460,00 kr
    3.460,00 kr
    1 * 1 Each
       3.460,00 kr
  • BS-10
    PFP-Biotin kit
    PFP-Biotin kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 2.600,00 kr
    2.600,00 kr
    1 * 1 Each
       2.600,00 kr
  • BS-11
    PEG₂-Iodoacetyl-Biotin kit
    PEG₂-Iodoacetyl-Biotin kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 4.410,00 kr
    4.410,00 kr
    1 * 1 Each
       4.410,00 kr
  • BS-12
    Iodoacetyl-LC-Biotin kit
    Iodoacetyl-LC-Biotin kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 3.460,00 kr
    3.460,00 kr
    1 * 1 Each
       3.460,00 kr
  • BS-13
    Biotin-PDA kit
    Biotin-PDA kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 3.460,00 kr
    3.460,00 kr
    1 * 1 Each
       3.460,00 kr
  • BS-14
    Biotin-BMCC kit
    Biotin-BMCC kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 4.090,00 kr
    4.090,00 kr
    1 * 1 Each
       4.090,00 kr
  • BS-16
    Biotin-PEG₂-Amine kit
    Biotin-PEG₂-Amine kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 4.320,00 kr
    4.320,00 kr
    1 * 1 Each
       4.320,00 kr
  • BS-17
    Biotin-PEG₃-LC-Amine kit
    Biotin-PEG₃-LC-Amine kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 5.940,00 kr
    5.940,00 kr
    1 * 1 Each
       5.940,00 kr
  • BS-18
    Biotin-Hydrazide kit
    Biotin-Hydrazide kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 2.510,00 kr
    2.510,00 kr
    1 * 1 Each
       2.510,00 kr
  • BS-19
    Biotin-LC-Hydrazide kit
    Biotin-LC-Hydrazide kit
    1 KIT
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 3.290,00 kr
    3.290,00 kr
    1 * 1 Each
       3.290,00 kr
Accessories
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • Optimizer Buffer™-V, 5X
    Optimizer Buffer™-V, 5X
    BKC-08
    2
    Log ind for tilgængelighed
    1 * 2 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • Optimizer Buffer™-III, 5X
    Optimizer Buffer™-III, 5X
    BKC-06
    2
    Log ind for tilgængelighed
    1 * 2 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • Optimizer Buffer™-IV, 5X
    Optimizer Buffer™-IV, 5X
    BKC-07
    2
    Log ind for tilgængelighed
    1 * 2 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • Optimizer Buffer™-VI, 5X
    Optimizer Buffer™-VI, 5X
    BKC-09
    2
    Log ind for tilgængelighed
    1 * 2 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • Optimizer Buffer™-II,5X
    Optimizer Buffer™-II,5X
    BKC-05
    2
    Log ind for tilgængelighed
    1 * 2 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Pakn
    Tilgængelighed
    Price
  • Optimizer Buffer™-I, 5X
    Optimizer Buffer™-I, 5X
    BKC-04
    2
    Log ind for tilgængelighed
    1 * 2 Items - 1.120,00 kr