Orders LinkContactUsLinkComponent
Protein Transfer buffers
Protein Transfer buffers
Western transfer buffer, Efficient™, 20X, 1 L
Katalog # 786-019
Leverandør:   G-Biosciences
Protein Transfer buffers
Katalog # 786-019
Leverandør:  G-Biosciences
CAS nummer:  
Protein Transfer buffers
Western transfer buffer, Efficient™, 20X, 1 L
CAS nummer:  
Price
841,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
Efficient™ Western Transfer Buffer (20X) achieves increased protein transfer efficiency. This ready-to-use transfer buffer is prepared for optimal conductivity and efficient protein transfer, without generating excessive heat or transfer distortion.
Specifikationer
Specifikationer
  • Pakn:
    1 L
  • Beskrivelse:
    Western transfer buffer, Efficient™, 20X