Section lifter
255 mm, 150 mm
Katalog # USBE3503
Leverandør: USBECK Laborgeräte
Gå til:
Product Details & Documents
Stainless steel blade, wooden handle. Section lifter with highly flexible blade and polished handle.
Specifikationer
- Længde:255 mm
- Klinge længde:150 mm