Disposable particle filtering respirators
Katalog # VALMVSPX353-07C2
Leverandør: DELTALYOVALMY
Specifikationer
- Model:Cup-shape
- Seq:02
- Pakn:10
- Klasse:FFP3
Specifikationer
Om denne vare
These comfortable disposable FFP2/FFP3 masks are made from 100% recyclable material and protects against odors caused by organic gases and acids.
- Airtightness is guaranteed thanks to its patented nose seal
- Low breathing resistance
- Less blur and perspiration
- Latex-, silicone- and PVC-free
Certifications: EN 149:2001+A1:2009