Microplate shakers, Jitterbug
Supplier: BOEKEL SCIENTIFIC
About this item
Designed to suitable for two or four standard microplates or strip trays.
- Calibration 'Mode'
- Temperature calibration
- Heated lid to provide greater uniformity
- Independently set temperature, shaking speed and mixing duration
2 Options Available Below
Product Details & Documents
Designed to suitable for two or four standard microplates or strip trays.
- Calibration 'Mode'
- Temperature calibration
- Heated lid to provide greater uniformity
- Independently set temperature, shaking speed and mixing duration
Specifications for Product Family
Specifications
- Catalog No:
- BOEI270400
- BOEI270440
- Model:
- Jitterbug-2
- Jitterbug-4
- Hastighed:
- 250 - 1200 min⁻¹
- 250 - 1200 min⁻¹
- Orbit:
- 2,0 mm
- 2,0 mm
- Plads til antal mikroplader:
- 2
- 4
- Vægt:
- 7,25 kg
- 7,25 kg
- B×D×H:
- 265×270×145 mm
- 385×270×145 mm
- Tidsindstilling:
- 1 min - 96 hrs
- 1 min - 96 hrs
Specifications
Product Family Options
Product Information
- BeskrivelseAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.BOEI270400ModelJitterbug-2Hastighed250 - 1200 min⁻¹Orbit2,0 mmPlads til antal mikroplader2Vægt7,25 kgB×D×H265×270×145 mmTidsindstilling1 min - 96 hrs - Specifications:
More
Specifications
Cat. No.BOEI270440ModelJitterbug-4Hastighed250 - 1200 min⁻¹Orbit2,0 mmPlads til antal mikroplader4Vægt7,25 kgB×D×H385×270×145 mmTidsindstilling1 min - 96 hrs