Casting tape, Scotchcast™ Plus
Casting tape, black, 3,6 m roll, 50 mm width
Katalog # MMMEM82002A
Leverandør: 3M MEDICAL
Gå til:
Product Details & Documents
This casting tape consists of a knitted fiberglass fabric impregnated with polyurethane resin. Exposure to moisture or water initiates a chemical reaction which causes the tape to become rigid.
- Finished casts produced from this casting tape are lightweight, strong, radiolucent, and porous
- Scotchcast™ Plus casting tape contains a lubricating agent in the resin to provide a slippery feel when exposed to water
Specifikationer
- Beskrivelse:Casting tape, black, 3,6 m roll, 50 mm width