VWR® Q Path® Safemount, Mounting Agents for Microscopy
Catalog # 00647520.
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pakning:Plastic bottle
- Pakn:500 mL
- Beskrivelse:Safemount mounting media Q Path®
Specifications
About this item
Safemount Q Path® is a mounting medium, supplied in an PET bottle, which is compatible with Safesolv. It glues the coverglass to the slide and fixes the probe in position after staining.
- Amber bottle
- Easily spreadable
- Easy to sponge away