Orders LinkContactUsLinkComponent
Chain clamp
Chain clamp
0 - 150 mm
Katalog # RLAA9100
Leverandør:   R L ENTERPRISES
Chain clamp
Katalog # RLAA9100
Leverandør:  R L ENTERPRISES
Chain clamp
0 - 150 mm
Price
1.210,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This rubber covered chain with cork lined jaws will support articles from 0 to 150 mm diameter.
  • Jaws are tightened by use of a plastic headed screw
  • The shaft and tensioner parts are nickel-plated
  • The shaft diameter is 10 mm
Specifikationer
  • Spændvidde:
    0 - 150 mm