Orders LinkContactUsLinkComponent
Filling System, RF-3
Filling System, RF-3
Owl™ RF-3 Filing system, 8 (2×4) compartments, 228,5×152,5×305 mm
Katalog # OWLARF-3
Leverandør:   THERMO OWL SCIENTIFIC
Filling System, RF-3
Katalog # OWLARF-3
Leverandør:  THERMO OWL SCIENTIFIC
Filling System, RF-3
Filling System, RF-3
Owl™ RF-3 Filing system, 8 (2×4) compartments, 228,5×152,5×305 mm
Price
2.210,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Organize your refrigerator, coldroom, freezer or benchtop with the Thermo Scientific™ Owl™ RF-3 Filing System.
  • Eight compartments constructed of durable 640 mm thick acrylic
  • No-skid rubber feet prevent sliding
  • Use either vertically or horizontally
Specifikationer
  • Beskrivelse:
    Owl™ RF-3 Filing system, 8 (2×4) compartments
  • B×D×H:
    228,5×152,5×305 mm