Cuvette for LKB analyser
Round cuvettes, 3,5 ml, PP, 12×51 mm
Katalog # SARS68.752
Marketsource
Leverandør: SARSTEDT
Gå til:
Product Details & Documents
Polypropylene round opaque cuvette for sample preparation with LKB analyser.
Specifikationer
- Diameter×H:12×51 mm
- Volumen:3,5 ml
- Beskrivelse:Round cuvettes
- Materiale:PP