ContactUsLinkComponent
HPLC columns, Eurospher I
 :  KNAUER
HPLC columns, Eurospher I
 :  KNAUER
  • Catalog No:
  • KNAU25DE181ESJ
  • KNAU25DE081ESJ
  • Kulstofbelastning:
  • 16%
  • 16%
  • Endcapped:
  • Ja
  • Ja
  • pH range:
  • 2 - 8
  • 2 - 8
  • Surface area:
  • 350 m²/g
  • 350 m²/g
  • USP code:
  • L1
  • L1
Frequently Bought Together