Blood collection set, Minivette®
Katalog # SARS17.2112.050
Leverandør: SARSTEDT
Specifikationer
- Volumen:50 µl
- Beskrivelse:Blood collection sets, Minivette®
- Pakn:2.000
Specifikationer
Om denne vare
Blood collection set for the precise dispensing of small volumes.
- Easy sample recovery
- Hygienic and secure sample transfer between patient and test equipment
Used in highly infectious nursing sectors.
Packaging: Individually wrapped.