Men's sweat jacket, Acode® Basecamp
Leverandør: FRISTADS KANSAS
Om denne vare
Double face sweat jacket made from 80% cotton and 20% polyester. Ideal for outdoor activities.
- Red contrast colour at collar and zip pullers
- Two front pockets, one brreast pocket with zip
- Cuffs and hem in rib with elastane
Weight: 320 g/m²
18 tilgængelige muligheder nedenfor
Product Details & Documents
Double face sweat jacket made from 80% cotton and 20% polyester. Ideal for outdoor activities.
- Red contrast colour at collar and zip pullers
- Two front pockets, one brreast pocket with zip
- Cuffs and hem in rib with elastane
Weight: 320 g/m²
Documents
Specifications for Product Family
Specifikationer
- Catalog No:
- KWIN110169543S
- KWIN110169543M
- KWIN110169543L
- KWIN110169543XL
- KWIN1101695432XL
- KWIN1101695433XL
- KWIN110169940S
- KWIN110169940M
- KWIN110169940L
- KWIN110169940XL
- KWIN1101699402XL
- KWIN1101699403XL
- KWIN110169941S
- KWIN110169941M
- KWIN110169941L
- KWIN110169941XL
- KWIN1101699412XL
- KWIN1101699413XL
- Outfit applikationer:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Risk area:
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Køn:
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- For mænd
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Lukkestørrelse:
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Med lynlås
- Stof:
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
- 80% cotton and 20% polyester
Specifikationer
Product Family Options
Produktinformation
- FarveTøj størrelseTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.KWIN110169543SOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN110169543MOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN110169543LOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN110169543XLOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN1101695432XLOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN1101695433XLOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN110169940SOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN110169940MOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN110169940LOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN110169940XLOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN1101699402XLOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN1101699403XLOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN110169941SOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN110169941MOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN110169941LOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN110169941XLOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN1101699412XLOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester - Specifikationer:
More
Specifications
Cat. No.KWIN1101699413XLOutfit applikationerGeneral PurposeRisk areaColdKønFor mændSleeve or leg lengthLongLukkestørrelseMed lynlåsStof80% cotton and 20% polyester