Analogue hygrometer
Carbide method (CM) hygrometer, 0 - 1,6 bar
Katalog # SIMELI0055
Leverandør: Industrial Physics
Gå til:
Product Details & Documents
This high precision destructive hygrometer is designed for applications in concrete, sand cement, and other construction materials. The water present in the sample chemically reacts with calcium carbide produces pressure in the vessel, which is proportional to the residual moisture content.
- Fast and reliable moisture determination
- Insensitive to external influences
- Standard splash- /dustproof
- Direct reading of the CM% moisture
- For printer documentation to protect against loss and follow-up costs
Delivery information: Supplied in carrying case with balance, pressure cylinder, calcium carbide and cleaning tools.
Documents
TQC Company Video
Videos
Specifikationer
- Område:0 - 1,6 bar
- Beskrivelse:Carbide method (CM) hygrometer